Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Cell Slide
Supplier: VWR
VWR®
CXD206010 CXD310010 CXD310008 CXD206008
76013-898CS 281.2 USD InStock
76013-898 76013-896 76013-894 76012-964
VWR® Cell Slide
Culture Dishes Cell Culture Dishes
The handle of the Cell Slide tilts, allowing operators the convience of direct clamping
  • Manufactured from unbreakable, high strength PET
  • Only the surface of the Cell Slide has been treated to promote cell adherence; other parts of the device are designed to deter cell adherence
  • 2 sizes are avialable: 8 mm and 10 mm
  • 12, 18, 32, or 45 Cell Slides are available on both the 8 mm and 10 mm models
  • High transparency and optimal light transmittance allow observation under a light microscope or fluorescence microscope
  • Set structure supports the development of multi-factor, multi-index, and multi-level in vitro study; resulting in accurate and reliable research results
DNase- and RNase-free. Non-pyrogenic.
The handle is also engraved with numbers to facilitate identification. The integrated Cell Slides are fixed in the culture dish, and if further testing is necessary the Cell Slides can be individually removed and placed in a 48-Well plate or similar container.
Certifications: ISO13485:2003 and ISO9001:2008.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty