Would you like to log in again?
Nick translation DNA labeling system 2.0, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN111-0050
76013-836EA 403.17 USD InStock
76013-836
Nick translation DNA labeling system 2.0, Enzo Life Sciences
Protein Linking and Labeling Systems
Nick translation system for preparing FISH probes
  • Ready-to-use Nick Translation Enzyme Mix minimizes pipetting error
  • Short labeling time - only 1 hour
  • Validated with fluorophore, biotin, and digoxigenin labeled dUTPs
The Nick Translation DNA Labeling System 2.0 provides a simple and efficient method for generating labeled DNA
The Nick Translation DNA Labeling System 2.0 can accommodate a wide range of fluorophore-labeled, biotin-labeled, and digoxigenin-labeled nucleotides. In addition to choice of label, the kit design allows the user to optimize incorporation and product size by adjusting the ratio of labeled-dUTP to dTTP. The ready-to-use NT Enzyme Mix is user friendly and minimizes error from pipetting.
Probes labeled by nick translation can be used in many different hybridization techniques including: in situ hybridization (ISH), fluorescent in situ hybridization (FISH), screening gene banks by colony or plaque hybridization, DNA or RNA transfer hybridization, and reassociation kinetic studies.
Contains NT Enzyme Mix 250 µL, Reaction Buffer 250 µL, dNTP Mix (dATP, dGTP, dCTP) 250 µL, dTTP 180 µL, Stop Buffer 250 µL, Nuclease-free Water 5 mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty