Would you like to log in again?
PLAQPRO® Lp-PLA2 Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT179-0200
76013-838EA 5630 USD InStock
76013-838
PLAQPRO® Lp-PLA2 Assay, Enzo Life Sciences
Assays Enzyme Assays
Enzyme activity kit to measure Lp-PLA2 activity in  serum samples
  • Cost-efficient alternative to competitor's Lp-PLA2 detection system
  • Wide analytical measurement range from 0 - 400.0 nmol/min/mL
  • Analytical sensitivity of 0.236 nmol/min/mL
  • High specificity and reproducibility, lot-after-lot
  • Ready-to-use calibration standards and QC controls are provided
  • Designed to run on automated clinical chemistry analyzers
For the measurement of enzyme activity of Lp-PLA2 (lipoprotein-associated phospholipase A2) in serum samples.££The PLAQPRO® Lp-PLA2 Assay measures the enzyme activity of Lp-PLA2 (lipoprotein-associated phospholipase A2) in serum samples. The assay is designed to run on automated clinical chemistry analyzers.
Contains Lp-PLA2 assay buffer, Lp-PLA2 assay substrate, 5 calibrator standards, QC low control, QC high control.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty