Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Digoxigenin IgG Polyclonal Antibody

Supplier: Enzo Life Sciences
ABS303-6000
76014-148EA 516 USD InStock
76014-148
Anti-Digoxigenin IgG Polyclonal Antibody
Antibodies
Recommended Applications: ISH (in situ hybridization)

Species reactivity: Species independent

Type: Primary
Antigen: Digoxigenin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty