Would you like to log in again?
CUVETTE SEMI MICRO 9 OPTICAL GLASS 0.7ML
Supplier: FIREFLYSCI, INC.
Semi-Micro Cuvette with PTFE Cover Type 9 5mm^ Semi-Micro Cuvette with PTFE Cover Type 9, Lightpath: 5mm, Material: Optical Glass, Outer Length: 7.5mm, Outer Width: 12.5mm, Outer Height: 45.0mm, Inner Length: 5mm, Inner Width: 4.0mm, Two windows optically polished, Capacity: 0.700ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CUVETTE SEMI MICRO 9 OPTICAL GLASS 0.7ML 76015-202 9G5 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty