Would you like to log in again?
CUVETTE SUB MICRO 701 UV QZ 8.5 MM 10 UL
Supplier: FIREFLYSCI, INC.
Sub-Micro Cuvette Type 701M 10mm^ Sub-Micro Cuvette Type 701M, Lightpath: 10mm, Material: Black UV Quartz, Outer Length: 12.5mm, Outer Width: 12.5mm, Outer Height: 45.0mm, Inner Length: 10mm, Inner Width: 1.0mm, z-Dimension: 8.5mm, Capacity: 10ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CUVETTE SUB MICRO 701 UV QZ 8.5 MM 10 UL 76016-112 701MUV10.10A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty