Would you like to log in again?
Macro Cuvettes, FireflySci
Supplier: FireFlySci
64FLUV10 62FLUV10 59FLUV10 53IR10 60G10 552UV10X4 606UV5B 64UV10 603FLUV10X2X2C 608FLUV1X1X11B 605UV1A 607IR3X3X43 601UV10A 64FLIR10 61IR0.5 514MUV10 552UV10X5 520MUV20 54FLIR10 520MUV5 51VUV10 520MIR20 606UV10A 61UV1 61UV10 58FLUV10 603FLUV3X3X11B 607UV3X3X43 520MUV10 605UV1B 601UV10B 59MIR10 61IR10 605UV2B 512UV 603FLUV1X1X11B 608FLUV3X3X11A 602UV10A 52G10X4 513MIR10 61FLUV10 601UV2B 62FLG10 601UV1A 606UV5A 54IR10 56IR10 61UV0.5 65FLUV10 521G100 521UV100 60UV10 53UV10 520MIR10 51VIR10 606UV10B 601UV5A 58UV10 61IR1 56UV10 6081.5X1.5X11B 552UV10X3 602UV10B 50UV10 52G10X1 605UV2A 52G10X5 520MIR5 509G10 58IR10 601UV5B 54FLUV10 603FL10X4X11B 514MIR10 62UV10 509UV10 526UV0.25 52G10X3 61FLIR10 51HUV10 510UV 529G5 5G10 601UV1B 607IR3X3X27 607UV3X3X27 601UV2A 552UV10X2 61UV5 61IR5 511UV 522G10 61IR2 62G10 64IR10 59MUV10 552UV10X1 52G10X2 50UV5 59FLIR10 5UV10 54UV10 608FLUV3X3X11B 603FLUV10X2X2B 608FLUV1X1X11A 61UV2 522UV10 515AUV10 6081.5X1.5X11A
76016-524EA 1751.11 USD InStock
76016-524 76016-496 76016-254 76016-252 76016-494 76016-258 76016-256 76016-498 76016-250 76016-492 76016-490 76016-416 76016-414 76016-418 76016-266 76016-420 76016-264 76016-302 76016-300 76016-422 76016-268 76016-262 76016-260 76015-782 76016-306 76016-428 76016-304 76016-426 76015-780 76016-308 76016-430 76016-276 76016-310 76016-274 76016-434 76016-278 76016-432 76016-272 76015-778 76016-270 76015-774 76015-776 76015-770 76015-772 76016-438 76016-436 76016-316 76016-442 76016-288 76016-286 76016-440 76016-446 76016-444 76016-280 76016-284 76015-768 76016-282 76015-766 76016-208 76016-448 76016-210 76016-298 76016-452 76016-296 76016-450 76016-456 76016-212 76016-290 76016-294 76016-292 76015-198 76016-218 76016-216 76016-458 76015-196 76016-464 76016-222 76016-462 76016-220 76016-500 76016-226 76016-468 76016-224 76016-466 76016-460 76016-504 76016-108 76016-502 76016-228 76016-508 76016-506 76016-474 76016-472 76016-110 76016-478 76016-512 76016-476 76016-510 76016-470 76016-516 76016-514 76016-518 76016-486 76016-484 76016-522 76016-520 76016-488 76016-482 76016-480
Macro Cuvettes, FireflySci
Cuvettes Spectrophotometer Cuvettes
Firefly's black quartz micro fluorescence flow cell.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in optical glass, IR quartz and UV quartz
These cuvettes are a good fit for many researchers with custom builds and machines. Highly accurate results, only need to test sample once. Firefly cuvettes are made with the best quality quartz and glass material available. Has excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a vigorous inspection process before leaving the factory.
Caution: Cuvettes must be scrupulously clean. Optical surfaces should not be touched as oil smudges are difficult to remove. Purity of solvents used for sample and cleaning is important. Only distilled water or spectrophotometric grade solvents should be used to avoid contamination from impure solvents. As soon as possible after use, cuvettes should be rinsed and soaked in distilled water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty