Would you like to log in again?
M6 Threaded Connectors for Tablet Dissolution Cells, A19
Supplier: FireFlySci
A19
76016-678EA 216.85 USD InStock
76016-678
M6 Threaded Connectors for Tablet Dissolution Cells, A19
Fittings and Connectors
A pair of M6 threaded connectors for use in Type 601, 602, and 606 tablet dissolution cells. The connector holds the PTFE tube firmly in place to seal it tightly against the quartz cell.
  • The dimensions of the supplied tubing is OD: 1.6 mm, ID: 0.8 mm. The length is 400 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty