Would you like to log in again?
Magnetic Centrifugal Stirrer for 10 mm Cells, FireflySci
Supplier: FireFlySci
P73
76016-692EA 42.42 USD InStock
76016-692
Magnetic Centrifugal Stirrer for 10 mm Cells, FireflySci
Stirrers Stirring Rods
This PTFE magnetic centrifugal stirrer fits into a standard 10 mm light path cuvette and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.
  • May also be used in test tubes
This one-piece molded unit will not scratch cuvettes and is equipped with crevices to trap dirt. Centrifugal pumping action generated by the cross channels in the upper face mixers without aeration. Photometric measurements can proceed uninterrupted while reagents are added.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty