Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UV/VIS Photometric Accuracy Calibration Standards, FireFlySci

Supplier: FireFlySci

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FUV-2.5 FUV-1.5 FUV-0.2 FUV-4.0 FUV-0.3 FUV-3.0 FUV-0.7
76016-708EA 2097.22 USD InStock
76016-708 76016-710 76016-724 76016-714 76016-722 76016-718 76016-726
UV/VIS Photometric Accuracy Calibration Standards, FireFlySci
Standards Color Reference and Spectroscopy Standards
Firefly's FUV neutral density solid-state filter series tests photometric accuracy in the UV and VIS range from 200-700nm. Our solid-state nano-deposition technology produces unparalleled stability and means that you will never need to recalibrate this filter.

  • Economic - no recalibration fees and replacement filters
  • Save time by having standards in your lab where they are needed
  • Save on having to handle fragile and toxic liquid calibration standards
  • Give yourself peace-of-mind knowing that all our standards are 100% NIST Traceable
  • High durability with scratch-resistant coating that protects optical surfaces

Capabilities: Photometric accuracy in the UV and VIS range
Stray light performance in the UV and VIS range (with matching HF Series filter)
These standards will work on any spectrophotometer.

Standard peaks included on certificate of calibration are 250, 280, 340, 360, 400, 465, 500, 546.1, 590 and 635nm.

Certifications: Fully NIST-traceable

Ordering information: For FUV filters that can do both photometric accuracy and stray light, please see the following link: FUV Dual Series.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty