Would you like to log in again?
LIGHT FILTER DUAL ABSORBANCE STRAY 2 AU
Supplier: FIREFLYSCI, INC.
FUV Dual Absorbance Stray Light Filter- 2.0au (1%T)Standard^ FUV-Series (200-700nm) Dual Absorbance Stray Light Filter- 2.0au (1%T)UV/VIS Photometric Accuracy & Stray Light Calibration Standard, Dimensions: 12.5x12.5x45 mm, NIST-Traceable filter that validates both stray light and absorbance
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIGHT FILTER DUAL ABSORBANCE STRAY 2 AU 76016-740 FUV-2.0D Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty