Would you like to log in again?
PHOTOMETRIC STANDARD HF-SRS 0.2 AU 45 MM
Supplier: FIREFLYSCI, INC.
HF-Series (400-700nm) Absorbance- 0.2au (60%T)Standard^ HF-Series (400-700nm) Absorbance- 0.2au (60%T)VIS Photometric Accuracy Calibration Standard, External dimensions: 12.5 x 12.5 x 45 mm, NIST-Traceable filter for validating photometric performance of a spectrophotometer.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PHOTOMETRIC STANDARD HF-SRS 0.2 AU 45 MM 76016-748 HF-0.2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty