Would you like to log in again?
PHOTOMETRIC STANDARD SPB SERIES 3AU 45MM
Supplier: FIREFLYSCI, INC.
SPB-Series (200-3300nm) Absorbance- 3.0au (0.1%T)Standard^ SPB-Series (200-3300nm) Absorbance- 3.0au (0.1%T) UV/VIS/NIR Photometric, Wavelength & Stray Light Calibration, External Dimensions: 12.5 x 12.5 x 45 mm, NIST-Traceable filter for validating photometric performance of a spectrophotometer.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PHOTOMETRIC STANDARD SPB SERIES 3AU 45MM 76016-798 SPB-3.0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty