Would you like to log in again?
KIT ULMT UV/VIS/NIR PHOTOMETRIC/LT CAL
Supplier: FIREFLYSCI, INC.
Ultimate UV/VIS/NIR Photometric, Wavelength & Stray Light Calibration Kit^ Superband UV/VIS/NIR Photometric Accuracy & Linearity Kit (10%T, 30%T, 90%T) (200-3000nm), External Dimensions: 12.5 x 12.5 x 45 mm, Tolerances: +/-0.0022 au, three filters and a blank
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT ULMT UV/VIS/NIR PHOTOMETRIC/LT CAL 76016-852 SPB-KIT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty