Would you like to log in again?
CABINET W/ 45 YLW BINS STEEL 18X36X78IN
Supplier: QUANTUM STORAGE SYSTEMS
Bin Cabinet Yellow 36 in^ Bin Locking Cabinet, Colour: Yellow, cabinet is made from 16 gauge steel and has a baked enamel finish, includes a 3-point locking system, with keys and has an all-welded construction for strength and durability, Number of Bins: 45, Outer Dimensions: 18 in x 36 in x 78 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET W/ 45 YLW BINS STEEL 18X36X78IN 76016-996 QPR-102YL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty