Would you like to log in again?
CABINET CLEARVIEW W/28 BL BINS STL 36IN
Supplier: QUANTUM STORAGE SYSTEMS
CLEAR-VIEW Security Bin Locking Cabinet Blue 36in^ CLEAR-VIEW Security Bin Cabinet, with Blue bins, features a clear door that provides visibility, Locking 14 Gauge Welded Gray Steel Cabinet With A 3-Point Locking Handle and Clear Door, With Two Keys No. Bins: 28, Outer Dimension: 36x18x78in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET CLEARVIEW W/28 BL BINS STL 36IN 76017-080 QSC-C240250BL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty