Would you like to log in again?
CABINET W/CLR BIN AND 240DRAWER STL 48IN
Supplier: QUANTUM STORAGE SYSTEMS
Storage Cabinet with Interlocking Drawer 48in^ Storage Cabinet with Interlocking Drawer, with Clear Bins, and 14 gauge all welded steel, Features, Gray baked-enamel finish, 3 point locking handle with 2 keys and with Louvered doors, No. Bins: 128, No. Drawers: 240, Outer Dimension: 48 x 24 x 78in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET W/CLR BIN AND 240DRAWER STL 48IN 76017-128 QSC-QIC161CL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty