Would you like to log in again?
CABINET W/56 BLACK BINS STEEL 36X24X72IN
Supplier: QUANTUM STORAGE SYSTEMS
Bin Cabinet Wide All-Welded Black bin 36in^ Bin Cabinet Wide All-Welded, with Black bin, Made from 14 gauge steel, with 3 point locking handle and two keys increase cabinets security, preassembled with an adjustable shelf, 12 door shelves, 4 drawers, Number of Bins: 58, Outer Dimension: 36x24x72in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET W/56 BLACK BINS STEEL 36X24X72IN 76017-174 QSC-3672-4DBK Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty