Would you like to log in again?
CABINET W/64 GREEN BINS STEEL 48X24X72IN
Supplier: QUANTUM STORAGE SYSTEMS
Bin Cabinet All-Welded with Green Bin 48in^ Bin Cabinet All-Welded with Green Bin, Made from 14in gauge all-welded steel for added strength, 2 adjustable interior shelves and also with three point locking handle and keys, with Gray baked enamel finish, No. Bins: 64, Outer Dimension: 48 x 24 x 72in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET W/64 GREEN BINS STEEL 48X24X72IN 76017-222 QSC-4805GN Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty