Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LIFT HYDRAULIC SSDPL-54-F 304 SS 750 LBS
Supplier: WESCO INDUSTRIAL PRODUCTS, LLC
Hydraulic Pedalift Jack^ Hydraulic Pedalift Jack, 304 SS, Four Wheeled, Model: SSDPL-54-F, 3inx18in forks sits at 5.75in, 8inx2in polyurethane tread non marking wheels with 440 SS sealed 3/4in precision ball bearing, 304 SS 3.5in polyurethane front swivel casters, 54in lift, Capacity: 750lbs
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIFT HYDRAULIC SSDPL-54-F 304 SS 750 LBS 76017-464 260197 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty