Would you like to log in again?
LIFT POWER FORK STACKER 64IN 2000 LBS
Supplier: WESCO INDUSTRIAL PRODUCTS, LLC
Powered Lift Fork Stacker Psfl-64-25-20S 2K^ Powered Lift Fork Stacker, Model:Psfl-64-25-20S 2K, 3inx25in forks, forks lower to floor, Adj. Fork: 6in - 24.5 OD, 64in raised height, 83 overall height, 20in leg ID, 26in leg OD, 15in load center, phenolic wheels and casters, 110V AC, Capacity: 2000 lbs
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIFT POWER FORK STACKER 64IN 2000 LBS 76017-630 261093 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty