Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Mini Electric Scissors Lift Table, MELT, Wesco International
Supplier: Wesco
270662
76017-666EA 5268.1 USD InStock
76017-666
Mini Electric Scissors Lift Table, MELT, Wesco International
Furniture Tables and Workbenches
Heavy duty to handle tough loads.
  • Small and light enough to move
  • Push button station
  • Load capacity 2200 lbs.
Full perimeter safety bar stops table from lowering if it contacts an obstructions.
Height: Raised 30"; Lowered 7"; Lift 30"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty