Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SlimT™ Mop Covers, Micronova

Supplier: Micronova
SlimT™
T6-10S TAA-12S T11-12S T11-18SIR T612-18S T20-14S T11-14S T20-18S TAA-14S T612-14S T6-12SIR TAW-18S T1112-18S T6-18S T6-14S T6-12S T11-18S TAA12-18S T11-14SIR TAA-18S T6-18SIR T612-18SIR
76017-990CS 260.99 USD InStock
76017-990 76017-980 76018-018 76017-992 76017-960 76017-994 76017-972 76017-984 76017-962 76018-072 76074-216 76017-952 76017-964 76017-954 76017-988 76017-956 76018-076 76018-022 76017-968 76018-000 76018-074 76017-948
SlimT™ Mop Covers, Micronova
Mops
Low-profile and economical, rounded T-mop covers that feature snaps on one side and a padded slip on the other to easily clean windows, awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialized cleaning.

  • Low profile, rounded covered sponges
  • Economical design for wide cleanroom use
  • Padded end protects equipment while cleaning
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces and under equipment
  • 12 and 18” options are ideal for use with the QPWT series Window Washing Tool
  • Holds up to strong disinfectants
  • Autoclavable

Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for removing residues and dispersing disinfectant
MegaTex™ - 100% non-woven polyamide is absorbent with a mildly abrasive surface
Microfiber - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry

The Micronova Slim T™ Mop Covers fit onto the 180° swivel T-Mop™ adapter. Available with an optional thicker foam layer to hold more disinfectant and facilitate wringing. The 30.5 cm (12”) and 45.7 cm (18”) options are designed for use with the Micronova Window Washing Tool for a streak-free finish.

Ordering information: All components must be ordered separately. For additional universal handle lengths, see universal mop handles from Micronova.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty