Would you like to log in again?
Label IT® MFP488 Nucleic Acid Labeling Kits
Supplier: Mirus Bio
Label IT®
MIR7125 MIR7100
76018-126EA 321.18 USD InStock
76018-126 76018-124
Label IT® MFP488 Nucleic Acid Labeling Kits
Transfection Reagents
Nucleic acid labeling reagents; label any DNA or RNA template – suitable for a range of applications.
  • High photostability – facilitates longer time for observation and image capture
  • Widespread Instrument compatibility – spectrally similar to fluorescein
  • Stable at Low pH – fluorescence is maintained over a broad range (pH 4 to 9)
  • One-step chemical method – easily and consistently control the labeling reactions
  • Adjustable labeling density – Achieve high sensitivity with optimally labeled DNA and RNA
  • Covalent mechanism – permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT® reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty