Would you like to log in again?
EZBio® Single-Use Assembly, Media Bottle, Polycarbonate, Vented with Tubing, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
438-0007-FLS 438-0004-FLS 438-0003-FLS 438-0006-FLS 438-0005-FLS
76018-358CS 1000.62 USD InStock
76018-358 76018-366 76018-364 76018-362 76018-360
EZBio® Single-Use Assembly, Media Bottle, Polycarbonate, Vented with Tubing, Sterile, Foxx Life Sciences
Bottles Plastic Bottles
EZBio® single-use polycarbonate bottle assemblies are an innovative and cost efficient option for single-use applications.
  • Perfect for fluid transfer and storage
  • VersaCap® technology
  • More cost efficient than stainless steel apparatus
  • Autoclavable
  • Sterilized
The EZBio® single-use sterile media bottles with tubing and vent filters are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components and Gamma Irradiated in the United States. Assemblies take advantage of Foxx’s VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate.
The caps for these bottles are vented with tubing, and sterilized.
Certifications: All materials comply with USP Class VI standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty