Would you like to log in again?
EZBio® Single-Use Media Bottle, Polycarbonate, Closed Cap, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
138-4211-FLS 138-5211-FLS 138-3211-FLS 138-2211-FLS 138-6511-FLS
76018-428PK 401.52 USD InStock
76018-428 76018-426 76018-424 76018-422 76018-420
EZBio® Single-Use Media Bottle, Polycarbonate, Closed Cap, Sterile, Foxx Life Sciences
Bottles Plastic Bottles
EZBio® single-use polycarbonate bottle assemblies are an innovative and cost efficient option for single-use applications.
  • Perfect for fluid transfer and storage
  • VersaCap® technology
  • More cost efficient than stainless steel apparatuses
  • Autoclavable
The EZbio® Single-Use Polycarbonate Media Bottles are perfect for fluid transfer and storage. Assemblies are made entirely with USP Class VI components. Assemblies take advantage of Foxx’s VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless steel apparatuses that are expensive to clean and validate.
They are autoclavable and made entirely with USP Class VI components.
Certifications: All materials comply with USP Class VI standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty