Would you like to log in again?
EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
25H-1707-FLS 25H-1702-FLS 25H-1703-FLS 25H-1705-FLS 25H-1701-FLS 25H-1708-FLS 25H-1706-FLS 25H-1704-FLS
76018-438EA 22.62 USD InStock
76018-438 76018-436 76018-434 76018-432 76018-444 76018-430 76018-442 76018-440
EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
Filters Venting Filters
EZBio® PTFE vent filters are used with EZBio® Single-Use bottle assemblies, which are perfect for fluid transfer and storage.
  • Vent filters used with EZBio® single-use bottle assemblies
  • PTFE membrane
  • Polypropylene housing
  • 0.22 µm
Certifications: All materials comply with USP Class VI standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty