Would you like to log in again?
Burned Fireclay Super Scorifiers, HarbisonWalker
Supplier: Harbison-Walker
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
141591 124371 124372
80701-894CS 1391.32 USD InStock
80701-894 11019-271 76018-764
Burned Fireclay Super Scorifiers, HarbisonWalker
Crucibles
Versatile in service, Super Scorifiers may be used to separate precious metals and ores from incinerated scrap samples.
  • Manufactured from high quality fireclay
  • Pressed to consistent dimensions and burned under closely controlled conditions
  • Manufacturing process results in more heats per scorifier, and excellent thermal shock resistance
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty