Would you like to log in again?
Tryptic Soy Broth, USP, 100 ml Bottle with Needle-Port Septum
Supplier: Hardy Diagnostics
U101
76018-776PK 322.05 USD InStock
76018-776
Tryptic Soy Broth, USP, 100 ml Bottle with Needle-Port Septum
Media for Microbiology Dehydrated Media for Microbiology
Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing.
  • Surface Sampling
  • Air Sampling
  • Sterility Tests
  • Quality culture media tested for performance, pH, and sterility
  • Kits are designed to help you follow USP <797> guidelines
  • Kits supply what is needed to test the proficiency and aseptic technique of technicians or pharmacists in a selfcontained ready-to-use format
  • Kits are available for low, medium, and high complexity procedures
Hardy Diagnostics offers all the products you need to easily assess the risk of microbial contamination of your CSPs (Compounded Sterile Preparations) according to USP Chapter <797>.
Certifications: Easily assess the risk of microbial contamination of CSPs (Compounded Sterile Preparations) according to USP Chapter 797.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty