Would you like to log in again?
Triple Polymer Cleanroom Glove, Mapa Professional
Supplier: Mapa Professionnel
517981
76018-960CS 145.73 USD InStock
76018-960
Triple Polymer Cleanroom Glove, Mapa Professional
Gloves
Tripolymer blend (nitrile, neoprene and natural rubber) outperforms natural rubber in applications involving aggressive chemicals.
  • Natural rubber protein levels test below detection
  • Strong and durable gloves are highly resistant to tears, snags and cuts
  • 20 mil thickness
  • Z-pattern exterior finish
  • Tapered flat cuffs with a raised diamond grip
Certifications: Finished, inspected, tested, and packaged in ISO Class 5 cleanroom.
Packaging: Pair-packaged in individual heat-sealed polybags.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty