Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safe-Lock Tubes, 2.0 ml - Microtubes

Supplier: Eppendorf
022363395 022363379 022363379 022363476 022363352 022363352 022363344 022600044 022363433 022363417 022363476 022363450 022363395 022363417 022363433 022600044 022363344 0030123620 022363450
20901-541CS 112.18 USD InStock
CA62111-754 CA20901-546 20901-541 CA20901-540 CA20901-541 20901-540 20901-543 20901-542 20901-545 CA20901-544 20901-544 CA20901-545 CA20901-542 20901-546 CA20901-543 CA53511-998 53511-998 76019-016 62111-754
Safe-Lock Tubes, 2.0 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock 2.0 ml microtube ensure secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2.0 ml capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.

  • Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss from tube breakage, and enhances safety when handling hazardous samples
  • Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimizes evaporation rates during prolonged storage
  • Large frosted lid and side surface for easy labeling
  • Functionality is assured within a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilization
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 2.0 ml microtubes redefine secure sample storage. The patented Safe-Lock mechanism ensures protection against accidental openings, preserving sample integrity meticulously. Versatile across laboratory applications, the 2.0 ml capacity adapts to diverse sample volumes. Constructed for durability, these microtubes withstand rigorous daily use. The user-friendly design, featuring clear graduations and a smooth surface, enhances visibility and handling ease. Elevate laboratory efficiency with Eppendorf® Safe-Lock tubes - trusted for reliability, security, and performance in sample management.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty