Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Compact Micro Centrifuges
Supplier: VWR
VWR®
2405-PCR 2418-37 2400-37 2405-MICRO-BC 2400-PCR 2405-MICRO 2400-MICRO-BC 2400-MICRO 2405-37
76019-206EA 5150.05 USD InStock
76019-206 10799-170 76019-208 76019-136 76019-138 76019-204 10799-122 76019-134 10799-174
VWR® Compact Micro Centrifuges
Centrifuges Microcentrifuges
These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.
  • Refrigerated Model with Pre-Cool Function Available
  • Robust and Steady Temperature Control
  • High Performance; Ideal for Sample Preparation
  • Short Run-up and Run-down Times
  • Pulse Button for Short Centrifugation Runs
Safety locking lids are equipped for biocontainment.
They are autoclavable and phenol-resistant. Operation noise made by these units is less than 58 dB, which is softer than a normal conversation.
Ordering information: Units come with a five-year manufacturer's warranty that extends the standard manufacturer's two-year warranty for an additional three years. The three-year extension is a full powertrain manufacturer's warranty on the centrifuge only. A comprehensive range of rotor options is available separately.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty