Would you like to log in again?
Neutralizing Buffered Peptone Water
Supplier: Hardy Diagnostics
C9243 C9242 K281 C9240 C9241 U183 U181
75878-366EA 1012.39 USD InStock
75878-366 75813-986 75838-644 76019-232 75838-646 76019-234 76019-230
Neutralizing Buffered Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
Used in the recovery of sub-lethally injured Salmonella species from industrial samples prior to selective enrichment and isolation.
This medium is recommended by the USDA FSIS (United States Department of Agriculture, Food Safety and Inspection Service) for poultry verification sampling.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty