Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® TPX™ Beakers with Handle

Supplier: VWR
VWR®
441060 441090 441040 441050 441080 441070
76019-280CS 48.99 USD InStock
76019-280 76019-286 76019-288 76019-278 76019-282 76019-284
VWR® TPX™ Beakers with Handle
Beakers
VWR® TPX™ Beakers, which are crystal-clear in color, have a handle, pouring spout and graduations printed directly on the beakers.

  • Chemical resistant
  • Integrated pouring lip
  • Autoclavable
  • Natural color
  • Printed graduation

The beakers are autoclavable and chemical resistant.

Working temperatures of 0 to 121 °C (32 to 248 °F) when used continuously.
Working temperatures as high as 170 °C (338 °F) when used for short periods of time.

Certifications: Resin meets US FDA 21 CFR requirements.

Caution: Do not stack products while autoclaving.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty