Would you like to log in again?
Impeller, PTFE Collapsible Blade, 1.5" Diameter, Caframo
Supplier: Caframo
A180 A180
76019-354EA 373 USD InStock
CA76019-354 76019-354
Impeller, PTFE Collapsible Blade, 1.5" Diameter, Caframo
Stirrers Stirring Blades
PTFE impeller for narrow neck vessels. PTFE blades attached to coated (5/16") diameter shaft. For high speed, low viscosity mixing.
  • PTFE blades – Excellent chemical (including acidic and caustic) resistance
  • Stainless steel shaft core – Provides stability to impeller
  • Scratch and chip resistant – Low risk of particles introduced to batch
A radial flow PTFE impeller for high speed, low viscosity mixing in narrow neck flasks. The blades are attached to an 8 mm (5/16") diameter PTFE-coated shaft, fanning out with agitation to a maximum diameter of 40 mm (1916"). The stainless steel core of the shaft provides stability. PTFE (polytetrafluoroethylene) can be used under the most adverse conditions. This high performance coating is insoluble, non-contaminating and has exceptional thermal resistance. It is easy to clean, does not scratch other surfaces and is unbreakable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty