Would you like to log in again?
Impeller, PTFE Four Blade Impeller, 1.5" Diameter, Caframo
Supplier: Caframo
A185 A185
76019-358EA 397.99 USD InStock
CA76019-358 76019-358
Impeller, PTFE Four Blade Impeller, 1.5" Diameter, Caframo
Stirrers Stirring Blades
Right hand rotation PTFE impeller with 45° pitched blades. Attached to coated 8 mm (5/16") diameter shaft. For high speed, low viscosity mixing.
  • PTFE blades, Excellent chemical (including acidic and caustic) resistance
  • Stainless steel shaft core, Provides stability to impeller
  • Scratch and chip resistant, Low risk of particles introduced to batch
  • Autoclavable and good for bio-tech processes
An axial flow PTFE impeller for high speed, low viscosity mixing. With 45° pitched blades, the 40 mm (19/16") diameter impeller is attached to an 8 mm (5/16") diameter PTFE-coated shaft. The stainless steel core of the shaft provides stability. PTFE (polytetrafluoroethylene) can be used under the most adverse conditions. This high performance coating is insoluble, non-contaminating and has exceptional thermal resistance. It is easy to clean, does not scratch other surfaces and is unbreakable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty