Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Samco™ Clicktainer™ Specimen Containers, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Samco™ Clicktainer™
120ORG53-1000 90WHT53-2002 90WHT53-1001 90WHT53-2001 90ORG53-1001 120WHT53-4000 90WHT53-1000 90WHT53-2000 90WHT53-2004 90WHT53-1002 120ORG53-4000 120WHT53-1000 90ORG53-1000 120WHT53-2000 90WHT53-2003 90GRN53-1000
76019-390CS 306.43 USD InStock
76019-390 76019-380 76019-374 76019-396 76019-386 76019-376 76019-388 76019-392 76019-370 76019-382 76019-384 76019-406 76019-400 76019-378 76019-402 76019-408
Samco™ Clicktainer™ Specimen Containers, Thermo Scientific
Sample Containers General Purpose Containers
Unique click seal system provides ultimate sample protection and user safety by ensuring correct application the first time.

  • Visual, audible, and tactile indication of correct cap closure
  • Wide 53 mm opening for ease of sample collection and management
  • Leakproof performance
  • 95 kPa compliant at ambient temperature
  • Ideal for use in pneumatic tube systems
  • Excellent clarity with easy to see graduations

Thermo Scientific understands the value of patient samples.

With this innovative closure system, Clicktainer™ Vials are ideally suited for the collection and transport of fluids, solids, and powders. They are perfect for a wide range of customer applications, from the collection of valuable and often hazardous patient samples such as urine, feces, and sputum and their transportation through hospital pneumatic/vacuum transport systems, to their suitability for use with formalin or fixatives for histology applications.

Choose safety first with the Samco™ Clicktainer™ Specimen Vials.

Sample collection needs to be easy. Sample transport needs to be secure. The Samco™ Clicktainer™ Vials have a wide opening and leakproof seal. The 'click' seal system indicates when the cap is properly applied so the sample is secure.

Certifications: CE marked in accordance with IVD Directicve 98/79/EC. Each batch pressure tested to 95kPa in accordance with BS EN 14401. Leak tested in accordance with BS EN 14254.

Caution: Note:  The term 'leakproof' applies to Thermo Scientific Samco™ Clicktainer™ products, 90 ml/53 mm and 120 ml/53 mm, that meet the following criteria: a) a vacuum test that draws 95 kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by one hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco™ Clicktainer™ Vials under conditions of their planned applications. Vials are leakproof at ambient temperature and pressure when used with their Clicktainer™ closures.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty