Would you like to log in again?
REMOVER IPA FLUX ISOCLEAN ESD SAFE 1GAL
Supplier: MicroCare
IsoClean* IPA-Based Flux Remover 1 Gal^ IsoClean* IPA-Based Flux Remover, Flash Point: 12 Deg C, Specific Gravity: 0.78, 0.79 at 25 Deg C, Viscosity: 2.43mPa/s, VOCs Content: 785g/L, Applications: Electronic Rework & Repair, For Microcare Trigger Grip Cleaning System, Size: 1 gal in mini pail
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER IPA FLUX ISOCLEAN ESD SAFE 1GAL 76019-892 MCC-BACG Case of 6 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty