Would you like to log in again?
REFILL WIPE PROCLEAN PRESAT FLUX REMOVER
Supplier: MicroCare
Proclean* Flux Remover Presaturated Wipe Refill^ Proclean* Flux Remover Presaturated Wipe, Refill, Blend Of Three Ultra-Pure Alcohols, Cleans R, Ra, Rma, Oa, In No-Cleanin Fluxes & Solder Paste, Static-Safe, Medium Drying, Moderate Evaporation, Mild Alcohol Odor, Excellent Toxicity, Biodegradable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REFILL WIPE PROCLEAN PRESAT FLUX REMOVER 76019-905 MCC-PROWR Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty