Would you like to log in again?
REMOVER LEAD-FREE FLUX POWERCLEAN 9X9IN
Supplier: MicroCare
PowerClean* Lead-Free Flux Remover 5 Gallon^ PowerClean* Lead-Free Flux Remover, Strong defluxer of high-temperature lead-free solder flux residues, Cleans oils, inks & conformal coatings, Eliminates white residues, Nonflammable & fast-drying, Removes acrylic conformal coatings, Volume: 5 Gallon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER LEAD-FREE FLUX POWERCLEAN 9X9IN 76019-910 MCC-PW2P Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty