Would you like to log in again?
WIPE UNCURED PRE-SATURATED EXPOXY VOC
Supplier: MicroCare
ExPoxy* Uncured Pre-Saturated Wipe^ ExPoxy* Uncured Epoxy Cleaner Pre-Saturated Wipe, Dissolves uncured chip bonder, epoxy glues, fluxes, pastes, grease and grime, 8x5in lint-free fabric, Unlimited shelf-life, Slow-drying, No residues, Plastic-safe, Ozone-safe, Low GWP, 100% VOC, Flammable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPE UNCURED PRE-SATURATED EXPOXY VOC 76019-917 MCC-EPXW Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty