Would you like to log in again?
REFILL WIPE EXPOXY UNCURED PRE-SATURATED
Supplier: MicroCare
ExPoxy* Uncured Pre-Saturated Wipe refill^ ExPoxy* Uncured Epoxy Cleaner Pre-Saturated Wipe refill, Dissolves uncured chip bonder, epoxy glues, fluxes, pastes, grease & grime, 8x5in lint-free fabric, Unlimited shelf-life, Slow-drying, No residue, Plastic-safe, Ozone-safe, Low GWP, 100%VOC, Flammable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REFILL WIPE EXPOXY UNCURED PRE-SATURATED 76019-919 MCC-EPXWR Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty