Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
REMOVER GENERAL PURPOSE FLUX 10.5 OZ
Supplier: MicroCare
General Purpose Flux Remover C 10.5oz^ General Purpose Flux Remover C, Odor: Slight, Ethereal, Boiling Point: 37 deg C (99 deg F), Use on SMT & BGA boards, hybrids, cables & connectors, Safe on plastics, Non-flammable, Designed for all types of electronics, SMT designs, BGA chips, Volume: 10.5 oz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER GENERAL PURPOSE FLUX 10.5 OZ 76019-922 MCC-FRC Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty