Would you like to log in again?
REMOVER GENERAL PURPOSE FLUX 1 GALLON
Supplier: MicroCare
General Purpose Flux Remover C 1 Gallon^ General Purpose Flux Remover C, Odor: Slight, Ethereal, Boiling Point: 37 deg C (99 deg F), Use on SMT & BGA boards, hybrids, cables & connectors, Safe on plastics, Non-flammable, Designed for all types of electronics, SMT designs, BGA chips, Volume: 1 Gallon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER GENERAL PURPOSE FLUX 1 GALLON 76019-924 MCC-FRCG Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty