Would you like to log in again?
Anti-Static Circuit Chiller & Freeze Spray, MicroCare
Supplier: MicroCare
MCC-FRZA
76019-930CS 757.9 USD InStock
76019-930
Anti-Static Circuit Chiller & Freeze Spray, MicroCare
Freezing Sprays
This anti-static circuit chiller freeze spray quickly finds circuit board faults.
  • No residue / no rinsing
  • Leaves ice on the PCB momentarily to freeze it
  • Dries instantly
  • Chills to lower than -45˚F/-43ºC in seconds; may go as low as -60˚F/-51˚C depending upon ambient conditions
  • Safe on plastics
  • Chemical Family: HFC blend
  • Nonflammable for better safety
This ultra-pure anti-static circuit chiller freeze spray uses an HFC propellant. So, it will not generate the component-shattering thermal shock that can occur with old-style HCFC-based chillers. ESD-safe when used in combination with the StatZap™ ESD Eliminator Tool.
Applications:
- Use for circuit test and repair
- Works on old and new equipment
Certifications: RoHS/WEEE/REACH/EPA SNAP compliant
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty