Would you like to log in again?
FastPROTEIN Blue, MP Biomedicals
Supplier: MP Biomedicals
116550500
76020-014EA 392.86 USD InStock
76020-014
FastPROTEIN Blue, MP Biomedicals
Protein Purification Protein Purification Kits
The FastPROTEIN blue kit is designed for rapid isolation and purification of protein from bacterial cultures.
  • Easy lysis of bacterial cells to release recombinant proteins
  • DNase and RNase-free lysing matrix tubes
The FastPROTEIN™ Blue Matrix, also known as Lysing Matrix B, is used with any FastPrep® Instrument to reproducibly lyse gram positive and gram negative bacterial cells within 40 seconds. Rapid release of recombinant protein saves valuable time during optimization of expression.
Information from up to 24 different induction variables can be quickly obtained after one 40 second FastPrep® run followed by electrophoresis. Once your protein of interest is expressed and identified, the active proteins released by the unique motion of the FastPrep® Instruments can be recovered from the lysate by a wide range of protein-specific techniques.
Application includes: Sample preparation; sample lysis; nucleic acid isolation and purification; molecular biology; gene expression; microbiological analysis; gene function; gene regulation.
Ordering information: FastPrep® Instrument or alternative high speed disruptor, empty lysing matrix tubes and caps, microcentrifuge, pipettmen, purification reagents.
Caution: Store at room temperature (15 - 30 °C)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty