Would you like to log in again?
DNA SET METHYLATED/NON-METHYLATED PUC19
Supplier: Zymo Research
Methylated & Non-Methylated pUC19 DNA Set^ Methylated & Non-Methylated pUC19 DNA Set, consists of two control DNAs (methylated and non-methylated) along with a set of specifically designed primers that can be used to assess the efficiency of bisulfite-mediated conversion of DNA
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DNA SET METHYLATED/NON-METHYLATED PUC19 76020-068 D5017 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty