Would you like to log in again?
HUMAN MATCHED DNA SET MALE HU 250 NG/UL
Supplier: Zymo Research
Human Matched DNA Set 250 ng/ul^ Human Matched DNA Set,  is a set of organ specific human genomic DNAs originating from single individual, ideal control for detection and quantification methods against 5-mC & 5-hmC, Concentration: 250ng/ul in modified TE buffer, Source: Human male, Storage </= -20C
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HUMAN MATCHED DNA SET MALE HU 250 NG/UL 76020-070 D5018 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty