Would you like to log in again?
REFILL WIPE ISOCLEAN IPA STENCIL 20X12CM
Supplier: MicroCare
IsoClean* Presaturated Wipe refill 20x12cm^ IsoClean* IPA Stencil Cleaner Presaturated Wipe in Plastic Tub, refill, for cleaning stencils, solder paste, fluxes, grease, light oils and general industrial grime, Noncorrosive & ESD-safe, 100% volatile, Flammable, Plastic-safe, Ozone-safe, Size: 20x12cm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REFILL WIPE ISOCLEAN IPA STENCIL 20X12CM 76020-143 MCC-BACWR Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty