Would you like to log in again?
CLEANER AQUEOUS STENCIL 1 GALLON CUBE
Supplier: MicroCare
Aqueous Stencil Cleaner 1 gallon cube^ Aqueous Stencil Cleaner, Cleans solder paste, uncured chip bonders & ink, Use on stencils, webbing, frames & misprinted boards, Fast cleaning, inside small stencil apertures, Nonflammable, noncorrosive, for ultrasonic cleaning tanks, Volume: 1 gallon cube
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANER AQUEOUS STENCIL 1 GALLON CUBE 76020-146 MCC-BGAG Case of 6 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty